Learn More
Medchemexpress LLC Apigenin-7-O-sulfate potassium | 120562-19-8 | 99.9% | 388.39 g·mol⁻1 | C15H9KO8S | 5mg
SDP
Catalog No. 5000270418
Encompass_Preferred
Apigenin-7-O-sulfate is a major flavonoid which is found to be a distinctive feature of plants from the Bay of Cadiz and can be extracted from these plants[1]
Catalog No. 50-002-70418 Supplier Medchemexpress LLC Supplier No. HYW7484565MG
Apigenin-7-O-sulfate is a major flavonoid which is found to be a distinctive feature of plants from the Bay of Cadiz and can be extracted from these plants[1]