Learn More
Selleck Chemical LLC Apigenin S2262-10mM/1mL
SDP
Catalog No. 503111772
Encompass_Preferred
Apigenin is a potent P450 inhibitor for CYP2C9 with Ki of 2 M
Catalog No. 50-311-1772 Supplier Selleck Chemical LLC Supplier No. S226210MM/1ML
Apigenin is a potent P450 inhibitor for CYP2C9 with Ki of 2 M