Learn More
Selleck Chemical LLC Apigenin S2262-1g
SDP
Catalog No. 503112810
Encompass_Preferred
Apigenin is a potent P450 inhibitor for CYP2C9 with Ki of 2 M
Catalog No. 50-311-2810 Supplier Selleck Chemical LLC Supplier No. S22621G
Apigenin is a potent P450 inhibitor for CYP2C9 with Ki of 2 M