Learn More
Abcam Apigenin, Anticancer agent, 10MG
Catalog No. 502707846
Encompass_Preferred
MW 270.24 Da, Purity >98%. Plant flavone. Antioxidant, anti-inflammatory and anticancer properties. Effects a wide range of molecular targets including caspases, fatty acid synthase, topoisomerase and nuclear factor-κB.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7846 Supplier Abcam Supplier No. AB12014110MG
MW 270.24 Da, Purity >98%. Plant flavone. Antioxidant, anti-inflammatory and anticancer properties. Effects a wide range of molecular targets including caspases, fatty acid synthase, topoisomerase and nuclear factor-κB.
The product is subject to the following: Abcam Restricted Use Statement