Learn More
Selleck Chemical LLC 4.7-Dimethoxy-5-Hydroxyflavone S3918-5mg
SDP
Catalog No. 503108844
Encompass_Preferred
4 7-Dimethoxy-5-Hydroxyflavone (4 7-dimethylapigenin apigenin dimethylether apigenin 7 4 -dimethyl ether) inhibits -glucosidase and -amylase enzymes and enhances 2-NBDG uptake in L6 cells
Catalog No. 50-310-8844 Supplier Selleck Chemical LLC Supplier No. S39185MG
4 7-Dimethoxy-5-Hydroxyflavone (4 7-dimethylapigenin apigenin dimethylether apigenin 7 4 -dimethyl ether) inhibits -glucosidase and -amylase enzymes and enhances 2-NBDG uptake in L6 cells