Learn More
Cayman Chemical Apigenin, Purity: ≥98%, CAS: 520-36-5, 50 mg
SDP
Catalog No. 502889578
Encompass_Preferred
Inhibits CK2 activity in the renal cortex with an IC50 value of 30 µM; potent inhibitor of NO and PGE2 biosynthesis by reducing iNOS and COX-2 expression
Catalog No. 50-288-9578 Supplier Cayman Chemical Supplier No. 1001027550 MG
Inhibits CK2 activity in the renal cortex with an IC50 value of 30 µM; potent inhibitor of NO and PGE2 biosynthesis by reducing iNOS and COX-2 expression