Learn More
Cayman Chemical Apigenin, Purity: ≥98%, CAS: 520-36-5, 100 mg
SDP
Catalog No. 502889655
Encompass_Preferred
Inhibits CK2 activity in the renal cortex with an IC50 value of 30 µM; potent inhibitor of NO and PGE2 biosynthesis by reducing iNOS and COX-2 expression
Catalog No. 50-288-9655 Supplier Cayman Chemical Supplier No. 10010275100 MG
Inhibits CK2 activity in the renal cortex with an IC50 value of 30 µM; potent inhibitor of NO and PGE2 biosynthesis by reducing iNOS and COX-2 expression