Learn More
Medchemexpress LLC Apigenin (Standard) | 520-36-5 | 270.24 | 100 MG
SDP
Catalog No. 5000416335
Encompass_Preferred
Apigenin (Standard) is the analytical standard of Apigenin, intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM.
  • Reference standard supplied for assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-16335 Supplier Medchemexpress LLC Supplier No. HYN1201R100MG
Apigenin (Standard) is the analytical standard of Apigenin, intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM.
  • Reference standard supplied for assay
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS