Learn More
Medchemexpress LLC Apigenin (Standard) | 520-36-5 | 270.24 | 25 MG
SDP
Catalog No. 5000389364
Encompass_Preferred
Apigenin (Standard) is the analytical standard of Apigenin, intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Reference standard supplied assay
  • For research use only
Catalog No. 50-003-89364 Supplier Medchemexpress LLC Supplier No. 25MGHYN1201R
Apigenin (Standard) is the analytical standard of Apigenin, intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Reference standard supplied assay
  • For research use only