Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apigenin (Standard) | 520-36-5 | 270.24 | 5 MG
SDP

Catalog No. 5000416336
Encompass_Preferred

Apigenin (Standard) is the analytical standard of Apigenin, intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. Features and benefits include:

  • Purity: 98.82%
  • Appearance: solid
  • Color: yellow to brown
  • Structure classification: flavonoids, phenols
  • Initial source: plants Compositae Baccharis sarothroides A.Gray, plants Umbelliferae
  • Endogenous metabolite: human gut microbiota metabolites

Catalog No. 50-004-16336 Supplier Medchemexpress LLC Supplier No. HYN1201R5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apigenin (Standard) is the analytical standard of Apigenin, intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM. This analytical standard is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS. Features and benefits include:

  • Purity: 98.82%
  • Appearance: solid
  • Color: yellow to brown
  • Structure classification: flavonoids, phenols
  • Initial source: plants Compositae Baccharis sarothroides A.Gray, plants Umbelliferae
  • Endogenous metabolite: human gut microbiota metabolites

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.