Learn More
Medchemexpress LLC Apigenin (standard) | 520-36-5 | 270.24 | 50 MG
SDP
Catalog No. 5000415970
Encompass_Preferred
Apigenin (Standard) is an analytical standard intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM and serves as a reference standard for assays.
  • Analytical standard for research and analytical applications
  • Competitive CYP2C9 inhibitor
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-15970 Supplier Medchemexpress LLC Supplier No. HYN1201R50MG
Apigenin (Standard) is an analytical standard intended for research and analytical applications. It is a competitive CYP2C9 inhibitor with a Ki of 2 μM and serves as a reference standard for assays.
  • Analytical standard for research and analytical applications
  • Competitive CYP2C9 inhibitor
  • Reference standard for assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications