Learn More
Medchemexpress LLC Apigenin (standard) | 520-36-5 | MFCD00006831 | 98.8% | 270.24 g/mol | C15H10O5 | 10 MG
SDP
Catalog No. 5000296179
Encompass_Preferred
Apigenin (4',5,7-trihydroxyflavone) analytical reference standard supplied as a purified solid for research and analytical applications. It is intended for qualitative and quantitative methods, including HPLC, GC, and MS, and is used as a calibration or assay reference (CAS 520-36-5).
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (~98.8%) for reliable calibration and assay performance.
  • Available in a 10 mg pack size for small-scale analytical workflows.
  • Solid form, stable under recommended storage conditions for reference use.
  • Suitable for method development, validation, and quantitative analysis.
Catalog No. 50-002-96179 Supplier Medchemexpress LLC Supplier No. HYN1201R10MG
Apigenin (4',5,7-trihydroxyflavone) analytical reference standard supplied as a purified solid for research and analytical applications. It is intended for qualitative and quantitative methods, including HPLC, GC, and MS, and is used as a calibration or assay reference (CAS 520-36-5).
  • Analytical standard suitable for HPLC, GC, and MS.
  • High purity (~98.8%) for reliable calibration and assay performance.
  • Available in a 10 mg pack size for small-scale analytical workflows.
  • Solid form, stable under recommended storage conditions for reference use.
  • Suitable for method development, validation, and quantitative analysis.