Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apigenin, Tocris Bioscience™

Catalog No. p-4783260
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-271-0 10 mg
12-275-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 12-271-0 Supplier R&D Systems Supplier No. 1227/10
This item is not returnable. View return policy

Protein kinase inhibitor

Protein kinase inhibitor. Suppresses tumor-promoting effects of TPA and exhibits antiproliferative activity in human breast cancer cells (IC50 values are 59.44 and 31.15 μM at 24 and 72 hrs respectively). Activates both the intrinsic and extrinsic apoptotic pathways and displays anti-inflammatory, hypotensive, antispasmodic and antioxidant properties in vivo.

Chemical Identifiers

CAS 520-36-5
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00006831
InChI Key KZNIFHPLKGYRTM-UHFFFAOYSA-N
Synonym apigenin, 5,7-dihydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, chamomile, spigenin, versulin, apigenol, 4',5,7-trihydroxyflavone, apigenine, c.i. natural yellow 1, 5,7,4'-trihydroxyflavone
PubChem CID 5280443
ChEBI CHEBI:18388
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
SMILES OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Quantity 10 mg
Formula Weight 270.2
Percent Purity >98%
Chemical Name or Material Apigenin
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.