Learn More
Medchemexpress LLC Apilimod | 541550-19-0 | 99.8% | 418.49 | 200 MG
SDP
Catalog No. 5000363102
Encompass_Preferred
Apilimod is a potent IL-12/IL-23 inhibitor, strongly inhibiting IL-12 with IC50s of 1 nM and 2 nM in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. It is also described as a potent and highly selective PIKfyve inhibitor.
  • Potent IL-12/IL-23 inhibitor
  • Strongly inhibits IL-12 with IC50s of 1 nM and 2 nM
  • Potent and highly selective PIKfyve inhibitor
  • Inhibits IFN-γ production in human PBMCs
  • Reduces severity of experimental autoimmune uveoretinitis (EAU)
  • Lowers level of IL-12 p40 in serum without altering body weight in EAU mice
  • Effective in Th1 model, with 51%±8% inhibition
Catalog No. 50-003-63102 Supplier Medchemexpress LLC Supplier No. HY14644200MG
Apilimod is a potent IL-12/IL-23 inhibitor, strongly inhibiting IL-12 with IC50s of 1 nM and 2 nM in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. It is also described as a potent and highly selective PIKfyve inhibitor.
  • Potent IL-12/IL-23 inhibitor
  • Strongly inhibits IL-12 with IC50s of 1 nM and 2 nM
  • Potent and highly selective PIKfyve inhibitor
  • Inhibits IFN-γ production in human PBMCs
  • Reduces severity of experimental autoimmune uveoretinitis (EAU)
  • Lowers level of IL-12 p40 in serum without altering body weight in EAU mice
  • Effective in Th1 model, with 51%±8% inhibition