Learn More
Medchemexpress LLC HY-14644 5mg Medchemexpress, Apilimod CAS:541550-19-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY146445MG
Encompass_Preferred
Medchemexpress, HY-14644 5mg Apilimod CAS:541550-19-0 Apilimod is a potent IL-12/IL-23 inhibitor, and strongly inhibits IL-12 with IC 50 s of 1 nM and 2 nM, in IFN-γ/SAC-stimulated human PBMCs and SAC-treated monkey PBMCs, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2704