Learn More
Selleck Chemical LLC Apilimod mesylate S0785-100MG
SDP
Catalog No. 5000041810
Encompass_Preferred
Apilimod mesylate is a potent and orally-available inhibitor of the cytokines interleukin-12 (IL-12) and interleukin-23 (IL-23) with the potential to treat certain autoimmune and inflammatory diseases Apilimod mesylate (STA-5326 mesylate) inhibits IL-12 with IC50 of 1 nM 1 nM and 2 nM in IFN- /SAC-stimulated human PBMCs human monocytes and mouse PBMCs respectively Apilimod mesylate (STA-5326 mesylate) is also a cell permeable small molecule that specifically inhibits PIKfyve with IC50 of 14 nM
Catalog No. 50-000-41810 Supplier Selleck Chemical LLC Supplier No. S0785100MG
Apilimod mesylate is a potent and orally-available inhibitor of the cytokines interleukin-12 (IL-12) and interleukin-23 (IL-23) with the potential to treat certain autoimmune and inflammatory diseases Apilimod mesylate (STA-5326 mesylate) inhibits IL-12 with IC50 of 1 nM 1 nM and 2 nM in IFN- /SAC-stimulated human PBMCs human monocytes and mouse PBMCs respectively Apilimod mesylate (STA-5326 mesylate) is also a cell permeable small molecule that specifically inhibits PIKfyve with IC50 of 14 nM