Learn More
TARGETMOL CHEMICALS INC APILIMOD MESYLATE 50MG
SDP
Catalog No. 502576923
Encompass_Preferred
Also available in 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apilimod mesylate (STA 5326 mesylate) is a potent IL-12/IL-23 inhibitor. purity: 99%
Catalog No. 50-257-6923 Supplier TARGETMOL CHEMICALS INC Supplier No. T353350MG
Also available in 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apilimod mesylate (STA 5326 mesylate) is a potent IL-12/IL-23 inhibitor. purity: 99%