Learn More
Medchemexpress LLC HY-14644A 5mg Medchemexpress, Apilimod (mesylate) CAS:870087-36-8 Purity:>98%
SDP
Catalog No. 501932705
Encompass_Preferred
Medchemexpress, HY-14644A 5mg Apilimod (mesylate) CAS:870087-36-8 Apilimod(STA 5326) mesylate is a potent IL-12/IL-23 inhibitor, IL-12 production in cultures of IFN-γ/LPS–stimulated human PBMCs is strongly inhibited by STA-5326 with an IC50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2705 Supplier Medchemexpress LLC Supplier No. HY14644A5MG
Medchemexpress, HY-14644A 5mg Apilimod (mesylate) CAS:870087-36-8 Apilimod(STA 5326) mesylate is a potent IL-12/IL-23 inhibitor, IL-12 production in cultures of IFN-γ/LPS–stimulated human PBMCs is strongly inhibited by STA-5326 with an IC50 of 10 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.