Learn More
Medchemexpress LLC Apilimod mesylate | 870087-36-8 | 99.8% | 610.70 | 50 MG
SDP
Catalog No. 5000360093
Encompass_Preferred
Apilimod mesylate is a chemical compound known for its potent inhibitory action against IL-12 and IL-23, with strong suppression of IL-12 at nanomolar concentrations. Additionally, it functions as a highly selective PIKfyve inhibitor, making it valuable for relevant biological research.
  • Potent IL-12/IL-23 inhibitor
  • Strongly inhibits IL-12 at nanomolar concentrations
  • Functions as a potent and highly selective PIKfyve inhibitor
  • Appears as a white to off-white solid
  • Inhibits IFN-γ production
  • Reduces serum IL-12 p40 levels in vivo
Catalog No. 50-003-60093 Supplier Medchemexpress LLC Supplier No. HY14644A50MG
Add to Cart
Add to Cart
Apilimod mesylate is a chemical compound known for its potent inhibitory action against IL-12 and IL-23, with strong suppression of IL-12 at nanomolar concentrations. Additionally, it functions as a highly selective PIKfyve inhibitor, making it valuable for relevant biological research.
  • Potent IL-12/IL-23 inhibitor
  • Strongly inhibits IL-12 at nanomolar concentrations
  • Functions as a potent and highly selective PIKfyve inhibitor
  • Appears as a white to off-white solid
  • Inhibits IFN-γ production
  • Reduces serum IL-12 p40 levels in vivo