Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apiopaeonoside | 100291-86-9 | 99.8% | 460.43 g/mol | C20H28O12 | 10 MG
SDP

Catalog No. 5000295674
Encompass_Preferred

Apiopaeonoside is a naturally occurring glycoside isolated from the root of Paeonia suffruticosa, provided as a research-grade reagent for biochemical and pharmacological studies. It has formula C20H28O12, a molecular weight of 460.43 g/mol, and is supplied in high purity for experimental applications.

  • High purity: 99.8%.
  • Molecular formula C20H28O12.
  • Molecular weight 460.43 g/mol.
  • Available as solid and as 10 mM solution in DMSO.
  • Intended for biochemical and pharmacological research use.

Catalog No. 50-002-95674 Supplier Medchemexpress LLC Supplier No. HYN216110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apiopaeonoside is a naturally occurring glycoside isolated from the root of Paeonia suffruticosa, provided as a research-grade reagent for biochemical and pharmacological studies. It has formula C20H28O12, a molecular weight of 460.43 g/mol, and is supplied in high purity for experimental applications.

  • High purity: 99.8%.
  • Molecular formula C20H28O12.
  • Molecular weight 460.43 g/mol.
  • Available as solid and as 10 mM solution in DMSO.
  • Intended for biochemical and pharmacological research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.