Learn More
Aviva Systems Biology Corp APIP Antibody - middle region (ARP56789_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56789_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human APIP. Alternate names: APIP2, CGI-29, MMRP19, dJ179L10.2, CGI29ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61989