Learn More
Cayman Chemical Apiin, Purity: ≥98%, CAS: 26544-34-3, 25 mg
SDP
Catalog No. 502948944
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; inhibits Aurora B kinase in a cell-free assay using the human enzyme (IC50 = 12.14 µM); inhibits LPS-induced increases in NO release and iNOS levels in J774.A1 macrophages at 10 and 50 µg/ml; scavenges DPPH, superoxide, and hydroxyl radicals (IC50s = 68, 390, and 48 µg/ml, respectively); increases SOD and catalase levels in mouse serum, brain, heart, liver, and kidney at 50 mg/kg
Catalog No. 50-294-8944 Supplier Cayman Chemical Supplier No. 3459625 MG
A flavonoid glycoside with diverse biological activities; inhibits Aurora B kinase in a cell-free assay using the human enzyme (IC50 = 12.14 µM); inhibits LPS-induced increases in NO release and iNOS levels in J774.A1 macrophages at 10 and 50 µg/ml; scavenges DPPH, superoxide, and hydroxyl radicals (IC50s = 68, 390, and 48 µg/ml, respectively); increases SOD and catalase levels in mouse serum, brain, heart, liver, and kidney at 50 mg/kg