Learn More
Medchemexpress LLC HY-50667 1g , Apixaban BMS-562247-01 CAS:503612-47-3 Purity:98%
SDP
Catalog No. 501492292
Encompass_Preferred
Medchemexpress, HY-50667 1g Apixaban BMS-562247-01 CAS:503612-47-3 Formula:C25H25N5O4 IC50: 0.08 nM (Human Factor Xa), 0.17 nM (Rabbit Factor Xa) Purity:98% Apixaban is a highly selective, reversible inhibitor of Factor Xa with K i of 0.08 nM and 0.17 nM in human and rabbit, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2292 Supplier Medchemexpress LLC Supplier No. HY506671G
Medchemexpress, HY-50667 1g Apixaban BMS-562247-01 CAS:503612-47-3 Formula:C25H25N5O4 IC50: 0.08 nM (Human Factor Xa), 0.17 nM (Rabbit Factor Xa) Purity:98% Apixaban is a highly selective, reversible inhibitor of Factor Xa with K i of 0.08 nM and 0.17 nM in human and rabbit, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.