Learn More
TARGETMOL CHEMICALS INC APIXABAN 1000MG
SDP
Catalog No. 502581104
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apixaban (BMS-562247-01) is an orally active inhibitor of coagulation factor Xa with anticoagulant activity. Apixaban directly inhibits factor Xa thereby interfering with the conversion of prothrombin to thrombin and preventing the formation of cross-linked fibrin clots. purity: 100%
Catalog No. 50-258-1104 Supplier TARGETMOL CHEMICALS INC Supplier No. T17361G
Also available in 2 mg 5 mg 10 mg 50 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Apixaban (BMS-562247-01) is an orally active inhibitor of coagulation factor Xa with anticoagulant activity. Apixaban directly inhibits factor Xa thereby interfering with the conversion of prothrombin to thrombin and preventing the formation of cross-linked fibrin clots. purity: 100%