Learn More
Medchemexpress LLC Apixaban-13C,d3 (BMS-562247-01-13C,d3) | 1261393-15-0 | 99.5% | C2413CH22D3N5O4 | 1 MG
SDP
Catalog No. 5000358949
Encompass_Preferred
Apixaban-13C,d3 is a deuterium and 13C labeled version of Apixaban, also known as BMS-562247-01-13C,d3. Apixaban is recognized as a highly selective and reversible inhibitor of Factor Xa, demonstrating Ki values of 0.08 nM in humans and 0.17 nM in rabbits.
  • Deuterium and 13C labeled compound.
  • Highly selective, reversible inhibitor of Factor Xa.
  • Can be used as a tracer in drug development processes.
  • Suitable as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.
Catalog No. 50-003-58949 Supplier Medchemexpress LLC Supplier No. HY50667S1MG
Apixaban-13C,d3 is a deuterium and 13C labeled version of Apixaban, also known as BMS-562247-01-13C,d3. Apixaban is recognized as a highly selective and reversible inhibitor of Factor Xa, demonstrating Ki values of 0.08 nM in humans and 0.17 nM in rabbits.
  • Deuterium and 13C labeled compound.
  • Highly selective, reversible inhibitor of Factor Xa.
  • Can be used as a tracer in drug development processes.
  • Suitable as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS.