Learn More
Medchemexpress LLC Apixaban | 503612-47-3 | MFCD11977295 | 99.99% | C25H25N5O4 | 200 MG
SDP
Catalog No. 5000357739
Encompass_Preferred
Apixaban is a highly selective, reversible, and orally active inhibitor of Factor Xa. It is used in the prevention and treatment of various thromboembolic diseases.
  • Highly selective and orally active factor Xa inhibitor
  • Reversible factor Xa inhibition
  • Ki values of 0.08 nM in human and 0.17 nM in rabbit
  • Used for prevention and treatment of thromboembolic disorders
Catalog No. 50-003-57739 Supplier Medchemexpress LLC Supplier No. HY50667200MG
Apixaban is a highly selective, reversible, and orally active inhibitor of Factor Xa. It is used in the prevention and treatment of various thromboembolic diseases.
  • Highly selective and orally active factor Xa inhibitor
  • Reversible factor Xa inhibition
  • Ki values of 0.08 nM in human and 0.17 nM in rabbit
  • Used for prevention and treatment of thromboembolic disorders