Learn More
Medchemexpress LLC HY-50667 50mg Medchemexpress, Apixaban CAS:503612-47-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5066750MG
Encompass_Preferred
Medchemexpress, HY-50667 50mg Apixaban CAS:503612-47-3 Apixaban (BMS-562247-01) is a highly selective, reversible and orally active inhibitor of Factor Xa with Ki of 0.08 nM and 0.17 nM in human and rabbit, respectively. Apixaban is in development for the prevention and treatment of various thromboembolic diseases[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9325