Learn More
Medchemexpress LLC Apixaban-¹³C,d₃ | 1261393-15-0 | 99.5% | C2413CH22D3N5O4 | 5 MG
SDP
Catalog No. 5000358274
Encompass_Preferred
Apixaban-13C,d3 is a deuterium and 13C labeled Apixaban, a highly selective, reversible inhibitor of Factor Xa. It is suitable for use as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. This product is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration may affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation in drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-58274 Supplier Medchemexpress LLC Supplier No. HY50667S5MG
Apixaban-13C,d3 is a deuterium and 13C labeled Apixaban, a highly selective, reversible inhibitor of Factor Xa. It is suitable for use as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. This product is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration may affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for quantitation in drug development
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs