Learn More
Medchemexpress LLC Apixaban-d3 | 1131996-12-7 | 99.9% | C25H22D3N5O4 | 5 MG
SDP
Catalog No. 5000357738
Encompass_Preferred
Apixaban-d3 (BMS-562247-01-d3) is a deuterium labeled version of Apixaban, a highly selective, reversible, and orally active inhibitor of Factor Xa. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs. This compound is suitable for use as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled Apixaban
  • Highly selective, reversible Factor Xa inhibitor
  • Useful as a tracer in drug development
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-57738 Supplier Medchemexpress LLC Supplier No. HY50667S15MG
Apixaban-d3 (BMS-562247-01-d3) is a deuterium labeled version of Apixaban, a highly selective, reversible, and orally active inhibitor of Factor Xa. Deuterium substitution can influence the pharmacokinetic and metabolic profiles of drugs. This compound is suitable for use as a tracer or an internal standard for quantitative analysis.
  • Deuterium labeled Apixaban
  • Highly selective, reversible Factor Xa inhibitor
  • Useful as a tracer in drug development
  • Can be used as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Potential to affect pharmacokinetic and metabolic profiles