Learn More
Selleck Chemical LLC Apixaban 50mg 503612-47-3 BMS 562247-01
SDP
Supplier:  Selleck Chemical LLC S159350MG
Encompass_Preferred
Apixaban (BMS 562247-01) is a highly selective, reversible inhibitor of Factor Xa with Ki of 0.08 nM and 0.17 nM in human and rabbit, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-580-1