Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apixaban impurity 4 | 27143-09-5 | C10H10Cl2N2O2 | 250 MG
SDP

Catalog No. 5000358477
Encompass_Preferred

Apixaban impurity 4 is an impurity of Apixaban, identified for laboratory use and substance manufacturing. This chemical presents as a white to off-white solid.

  • Molecular weight: 261.11
  • Molecular formula: C10H10Cl2N2O2
  • CAS number: 27143-09-5
  • Melting point: 147.5-149.5 °C
  • Boiling point: 337.1±44.0 °C at 760 mmHg
  • Stable under recommended storage conditions

Catalog No. 50-003-58477 Supplier Medchemexpress LLC Supplier No. 250MGHY77967
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apixaban impurity 4 is an impurity of Apixaban, identified for laboratory use and substance manufacturing. This chemical presents as a white to off-white solid.

  • Molecular weight: 261.11
  • Molecular formula: C10H10Cl2N2O2
  • CAS number: 27143-09-5
  • Melting point: 147.5-149.5 °C
  • Boiling point: 337.1±44.0 °C at 760 mmHg
  • Stable under recommended storage conditions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.