Learn More
Cayman Chemical ApIxabn-13C-d3 500ug
SDP
Supplier:  Cayman Chemical 25034500 UG
Encompass_Preferred
An internal standard for the quantification of apixaban by GC- or LC-MS
Catalog No. 50-293-4563