Learn More
Cayman Chemical Apixaban-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1261393-15-0, 5 mg
SDP
Catalog No. 502935220
Encompass_Preferred
An internal standard for the quantification of apixaban by GC- or LC-MS
Catalog No. 50-293-5220 Supplier Cayman Chemical Supplier No. 250345 MG
An internal standard for the quantification of apixaban by GC- or LC-MS