Learn More
Eurofins Discoverx Ready-to-Assay APJ Apelin Receptor Frozen Cells, 2 Vials of Cells. (HTS068RTA)
Catalog No. 502005795
Encompass
The Ready-to-Assay APJ Apelin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. apelin) based activation of AGTRL1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses AGTRL1 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of AGTRL1, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.
Catalog No. 50-200-5795 Supplier Eurofins Discoverx Supplier No. HTS068RTA
The Ready-to-Assay APJ Apelin Receptor Frozen Cells are a simple, thaw-and-assay cell format for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This assay assesses ligand (e.g. apelin) based activation of AGTRL1 GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses AGTRL1 alongside exogenous levels of a promiscuous G-protein, coupling the receptor to the calcium signaling pathway. Upon activation of AGTRL1, the resulting calcium activity can be fluorescently assayed. Ready-to-assay kits consist of two vials of the stably modified clonal cell line in a convenient thaw-and-use format. For detailed product information, User manual and datasheet, visit www.discoverx.com/products-applications/frozen-cells.