Learn More
Medchemexpress LLC APL180 TFA (L-4F TFA) | 98.8% | 2310.60 (free base) | 25 MG
SDP
Catalog No. 5000400381
Encompass_Preferred
APL180 TFA (L-4F) is an apolipoprotein AI mimetic peptide that enhances the anti-inflammatory activity of high-density lipoprotein (HDL). APL180 can be used in the study of cardiovascular diseases.
  • Apolipoprotein AI mimetic peptide
  • Enhances the anti-inflammatory activity of high-density lipoprotein (HDL)
  • Can be used in the study of cardiovascular diseases
Catalog No. 50-004-00381 Supplier Medchemexpress LLC Supplier No. 25MGHYP6365
APL180 TFA (L-4F) is an apolipoprotein AI mimetic peptide that enhances the anti-inflammatory activity of high-density lipoprotein (HDL). APL180 can be used in the study of cardiovascular diseases.
  • Apolipoprotein AI mimetic peptide
  • Enhances the anti-inflammatory activity of high-density lipoprotein (HDL)
  • Can be used in the study of cardiovascular diseases