Learn More
Medchemexpress LLC Aplaviroc hydrochloride | 461023-63-2 | 99.9% | 614.17 | 50 MG
SDP
Catalog No. 5000386428
Encompass_Preferred
Aplaviroc hydrochloride (AK 602) is supplied as a 50 MG quantity of a potent CCR5 antagonist. Derived from SDP, it is effective against various HIV-1 strains (HIV-1Ba-L, HIV-1JRFL, and HIV-1MOKW) with IC50s ranging from 0.1-0.4 nM. This product is intended for research use only and not for sale to patients.
  • Potent CCR5 antagonist
  • Active against multiple HIV-1 strains
  • Suitable for research applications
Catalog No. 50-003-86428 Supplier Medchemexpress LLC Supplier No. HY17450A50MG
Aplaviroc hydrochloride (AK 602) is supplied as a 50 MG quantity of a potent CCR5 antagonist. Derived from SDP, it is effective against various HIV-1 strains (HIV-1Ba-L, HIV-1JRFL, and HIV-1MOKW) with IC50s ranging from 0.1-0.4 nM. This product is intended for research use only and not for sale to patients.
  • Potent CCR5 antagonist
  • Active against multiple HIV-1 strains
  • Suitable for research applications