Learn More
Aviva Systems Biology Corp APLNR Antibody - N-terminal region (ARP59985_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59985_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APLNR. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human APLNR. Alternate names: AGTRL1, APJ, APJR, FLJ90771, FLJ96609, HG11, MGC45246ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-906