Learn More
Miele, Inc. APLW 001load carrier
Catalog No. 501953624
Encompass_Preferred
APLW 001 Load carrierFor laboratory glassware, e.g. beakersUseable surface: 490 x 540 mm
Catalog No. 50-195-3624 Supplier Miele, Inc. Supplier No. 11104480
APLW 001 Load carrierFor laboratory glassware, e.g. beakersUseable surface: 490 x 540 mm