Learn More
Miele, Inc. APLW 002load carrier
Catalog No. 501953625
Encompass_Preferred
APLW 002 Load carrierFor bottles (5 and 10 l), large Erlenmeyer flasks- Up to 4 items, ø max mm 240- Up to 5 items, ø max mm 190
Catalog No. 50-195-3625 Supplier Miele, Inc. Supplier No. 11104490
APLW 002 Load carrierFor bottles (5 and 10 l), large Erlenmeyer flasks- Up to 4 items, ø max mm 240- Up to 5 items, ø max mm 190