Learn More
Miele, Inc. APLW 011upper basket
Catalog No. 501953634
Encompass_Preferred
APLW 011 Upper basket- 110 injectors- Ø max. 40 mm- only for 2.5 mm Ø nozzles
Catalog No. 50-195-3634 Supplier Miele, Inc. Supplier No. 11104580
APLW 011 Upper basket- 110 injectors- Ø max. 40 mm- only for 2.5 mm Ø nozzles