Learn More
Miele, Inc. APLW 012upper basket
Catalog No. 501953635
Encompass_Preferred
APLW 012 Upper basketUpper basket- 84 injectors- Ø max. 35 mm- only for 2.5 mm Ø nozzles
Catalog No. 50-195-3635 Supplier Miele, Inc. Supplier No. 11104590
APLW 012 Upper basketUpper basket- 84 injectors- Ø max. 35 mm- only for 2.5 mm Ø nozzles