Learn More
Miele, Inc. APLW 013/1upper basket
Catalog No. 501953637
Encompass_Preferred
APLW 013/1 Upper basket- 42 injectors- Ø max. 70 mm- Max. height: 160 mm- incl. nozzles 42x APLW 051
Catalog No. 50-195-3637 Supplier Miele, Inc. Supplier No. 11104610
APLW 013/1 Upper basket- 42 injectors- Ø max. 70 mm- Max. height: 160 mm- incl. nozzles 42x APLW 051