Learn More
Miele, Inc. APLW 013/2upper basket
Catalog No. 501953638
Encompass_Preferred
APLW 013/2 Upper basket- 10 + 32 injectors- Ø max. 70 mm- Height max. 200 – 300 mm- incl. nozzles 32x APLW 051, 10x APLW 055
Catalog No. 50-195-3638 Supplier Miele, Inc. Supplier No. 11104620
APLW 013/2 Upper basket- 10 + 32 injectors- Ø max. 70 mm- Height max. 200 – 300 mm- incl. nozzles 32x APLW 051, 10x APLW 055