Learn More
Miele, Inc. APLW 018/1upper basket
Catalog No. 501953645
Encompass_Preferred
APLW 018/1 Upper basket- 24 + 121 injectors- Ø max. 20 + 75 mm- Max. height: 160 mm- incl. nozzles 121x APLW 048, 24x APLW 051
Catalog No. 50-195-3645 Supplier Miele, Inc. Supplier No. 11104690
Add to Cart
Add to Cart
Request a Quote
APLW 018/1 Upper basket- 24 + 121 injectors- Ø max. 20 + 75 mm- Max. height: 160 mm- incl. nozzles 121x APLW 048, 24x APLW 051