Learn More
Miele, Inc. APLW 019/1upper basket
Catalog No. 501953647
Encompass_Preferred
APLW 019/1 Upper basket- 121 injectors- Ø max. 25 mm- Max. height: 90 mm- incl. nozzles 121x APLW 047
Catalog No. 50-195-3647 Supplier Miele, Inc. Supplier No. 11104710
Add to Cart
Add to Cart
Request a Quote
APLW 019/1 Upper basket- 121 injectors- Ø max. 25 mm- Max. height: 90 mm- incl. nozzles 121x APLW 047