Learn More
Miele, Inc. APLW 020/1lower basket
Catalog No. 501953649
Encompass_Preferred
APLW 020/1 Lower basket- 121 injectors- Ø max. 25 mm- Max. height: 140 mm- incl. nozzles 121x APLW 047
Catalog No. 50-195-3649 Supplier Miele, Inc. Supplier No. 11104730
APLW 020/1 Lower basket- 121 injectors- Ø max. 25 mm- Max. height: 140 mm- incl. nozzles 121x APLW 047