Learn More
Miele, Inc. APLW 022lower basket
Catalog No. 501953651
Encompass_Preferred
APLW 022 Lower basket- 70 injectors- Ø max. 52 mm
Catalog No. 50-195-3651 Supplier Miele, Inc. Supplier No. 11104750
APLW 022 Lower basket- 70 injectors- Ø max. 52 mm