Learn More
Miele, Inc. APLW 024/1lower basket
Catalog No. 501953654
Encompass_Preferred
APLW 024/1 Lower basket- 42 injectors- Ø max. 70 mm- Max. height: 230 mm- incl. nozzles 42x APLW 052
Catalog No. 50-195-3654 Supplier Miele, Inc. Supplier No. 11104780
Add to Cart
Add to Cart
APLW 024/1 Lower basket- 42 injectors- Ø max. 70 mm- Max. height: 230 mm- incl. nozzles 42x APLW 052