Learn More
Miele, Inc. APLW 024/2lower basket
Catalog No. 501953655
Encompass_Preferred
APLW 024/2 Lower basket- 42 injectors- Ø max. 70 mm- Height max.: 200–300 mm- incl. nozzles 10x APLW 055, 32x APLW 051
Catalog No. 50-195-3655 Supplier Miele, Inc. Supplier No. 11104790
APLW 024/2 Lower basket- 42 injectors- Ø max. 70 mm- Height max.: 200–300 mm- incl. nozzles 10x APLW 055, 32x APLW 051