Learn More
Miele, Inc. APLW 025/1lower basket
Catalog No. 501953658
Encompass_Preferred
APLW 025/1 Lower basket- 24 injectors- Ø max. 70 mm- Height max.: 200–300 mm- incl. nozzles 12x APLW 056, 12x APLW 052
Catalog No. 50-195-3658 Supplier Miele, Inc. Supplier No. 11104820
APLW 025/1 Lower basket- 24 injectors- Ø max. 70 mm- Height max.: 200–300 mm- incl. nozzles 12x APLW 056, 12x APLW 052